Learn More
Seracare Life Sciences Inc TMB MEMBRANE HRP SUBSTRATE
Catalog No. 5067423
Encompass_Preferred
Most sensitive chromogenic substrate for peroxidase detection. It develops a deep blue color at the reaction site in the presence of peroxidase-labeled conjugates. Recommended for membrane assays and dot blots. 3-component format of 3, 3', 5, 5' - Tetramethylbenzidine, simply mix and use.
This item is not returnable. View return policy
Catalog No. 50-674-23 Supplier Seracare Life Sciences Inc Supplier No. 54200025
Most sensitive chromogenic substrate for peroxidase detection. It develops a deep blue color at the reaction site in the presence of peroxidase-labeled conjugates. Recommended for membrane assays and dot blots. 3-component format of 3, 3', 5, 5' - Tetramethylbenzidine, simply mix and use.