Learn More
Medchemexpress LLC HY-15930C 100mg Medchemexpress, TMB (monosulfate) CAS:54827-18-8 Purity:>98%
SDP
Catalog No. 501969963
Encompass_Preferred
Medchemexpress, HY-15930C 100mg TMB (monosulfate) CAS:54827-18-8 TMB monosulfate is a chromogenic substrate used in staining procedures in immunohistochemistry as well as being a visualizing reagent used in enzyme-linked immunosorbent assays (ELISA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9963 Supplier Medchemexpress LLC Supplier No. HY15930C100MG
Medchemexpress, HY-15930C 100mg TMB (monosulfate) CAS:54827-18-8 TMB monosulfate is a chromogenic substrate used in staining procedures in immunohistochemistry as well as being a visualizing reagent used in enzyme-linked immunosorbent assays (ELISA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.