Learn More
Surmodics Inc TMB One Component HRP Membrane Substrate, 100mL
SDP
Catalog No. NC1462819
Encompass_Preferred
The BioFX™ one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution for membrane applications. The staining pattern provides a dark blue precipitate with low background to increase the signal-to-noise ratios in western blot, dot blots and line assays
Catalog No. NC1462819 Supplier Surmodics Inc Supplier No. TMBM010001
The BioFX™ one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution for membrane applications. The staining pattern provides a dark blue precipitate with low background to increase the signal-to-noise ratios in western blot, dot blots and line assays