Learn More
Carbosynth LLC. High sensitivity TMB Peroxidase Substrate (stable stop) slow acting, ready to use in immunoassays
SDP
Catalog No. 501253316
Encompass_Preferred
This substrate produces a soluble end product which is blue in color that is quantitated at 650 nm. Use of a stop solution enhances sensitivity 2-4 fold and the resulting yellow solution can be read at 450 nm.
Catalog No. 50-125-3316 Supplier Carbosynth LLC. Supplier No. 42RTB1060.5L
This substrate produces a soluble end product which is blue in color that is quantitated at 650 nm. Use of a stop solution enhances sensitivity 2-4 fold and the resulting yellow solution can be read at 450 nm.