Learn More
Echelon Biosciences Research Labs TMB PRECIPITATING 22MLTMB PRE
SDP
Catalog No. 5082226
Encompass
TMB Precipitating (K-TMBP) is a chromogen substrate that reacts with horseradish peroxide (HRP) producing a permanent purple colored spot or band. This detection reagent is best used with an HRP conjugated secondary or tertiary antibody on a dot blot or w This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-822-26 Supplier Echelon Biosciences Research Labs Supplier No. KTMBP22ML
TMB Precipitating (K-TMBP) is a chromogen substrate that reacts with horseradish peroxide (HRP) producing a permanent purple colored spot or band. This detection reagent is best used with an HRP conjugated secondary or tertiary antibody on a dot blot or w This item is non returnable