Learn More
Crescent Chemical Co Inc TMB READY TO USE ELISA SUBS.
SDP
Supplier:  Crescent Chemical Co Inc SE37068.01
Encompass_Preferred
TMB ready to use ELISA substrate One bottle reagent: contains 3,3',5,5'-tetramethylbenzidine, buffer and peroxide in a single convenient, ready-to-use solution recommended for the detection of horseradish peroxidase in ELISA assays. Develops a deep blue colour that turns bright
Catalog No. 50-248-219