Learn More
Scytek Laboratories TMB Soluble Reagent (Standard Sensitivity)
SDP
Catalog No. NC1881229
Encompass_Preferred
This standard sensitivity liquid substrate for peroxidase consists of Tetramethylbenzidine (TMB) plus dilute hydrogen peroxide in a single-reagent stabilized form. The reagent has been specifically formulated for measuring peroxidase in ELISA systems which is stable for long-term storage providing sensitivity greater than OPD. This reagent is available as a single component, ready-to-use reagent. When used in conjunction with our proprietary TMB Stop Buffer (TSB), the resulting color product will not fade or darken for several hours.
Catalog No. NC1881229 Supplier Scytek Laboratories Supplier No. TM1999
This standard sensitivity liquid substrate for peroxidase consists of Tetramethylbenzidine (TMB) plus dilute hydrogen peroxide in a single-reagent stabilized form. The reagent has been specifically formulated for measuring peroxidase in ELISA systems which is stable for long-term storage providing sensitivity greater than OPD. This reagent is available as a single component, ready-to-use reagent. When used in conjunction with our proprietary TMB Stop Buffer (TSB), the resulting color product will not fade or darken for several hours.