Learn More
Seracare Life Sciences Inc TMB STOP SOLUTION 4X100ML CS
Catalog No. 5067443
Encompass_Preferred
Specifically designed for use with SureBlue Reserve TMB, SureBlue TMB, and TMB 2-Component Microwell Peroxidase Substrate product lines to stop color development in ELISA. Upon addition of stop solution, TMB turns yellow and can be read at 450 nm.
This item is not returnable. View return policy
Catalog No. 50-674-43 Supplier Seracare Life Sciences Inc Supplier No. 51500020
Specifically designed for use with SureBlue Reserve TMB, SureBlue TMB, and TMB 2-Component Microwell Peroxidase Substrate product lines to stop color development in ELISA. Upon addition of stop solution, TMB turns yellow and can be read at 450 nm.