Learn More
Enzo Life Sciences TMB substrate, (50 ml), (50ml), CAS Number: 54827-17-7
SDP
Catalog No. 50658086
Encompass_Preferred
This TMB Substrate is intended for use as a colorimetric HRP-substrate for immunoassay development. Each bottle is sufficient for use with 5x96 wells microtiter plates. Additional assay development reagents are available as companion products to our IMMUNOSET products. Alternative Name: 3,3',5,5'-Tetramethylbenzidine. CAS Number: 54827-17-7. Long Term Storage: +4°C
Catalog No. 50-658-086 Supplier Enzo Life Sciences Supplier No. ADI801805
This TMB Substrate is intended for use as a colorimetric HRP-substrate for immunoassay development. Each bottle is sufficient for use with 5x96 wells microtiter plates. Additional assay development reagents are available as companion products to our IMMUNOSET products. Alternative Name: 3,3',5,5'-Tetramethylbenzidine. CAS Number: 54827-17-7. Long Term Storage: +4°C