Learn More
Vector Laboratories TMB Substrate (Super Sensitive) - LS-M36, 100ml
SDP
Catalog No. NC3404480
Encompass
TMB Substrate (Super Sensitive) is a ready-to-use TMB substrate for microwell applications containing 3,3',5,5'-tetramethylbenzidine in a mildly acidic buffer. This substrate is our most sensitive TMB buffer. This sensitive, faster kinetic TMB substrate is approximately 40-fold more sensitive than our most popular substrate, TMB Substrate (ELISA).
Catalog No. NC3404480 Supplier Vector Laboratories Supplier No. LSM36100
TMB Substrate (Super Sensitive) is a ready-to-use TMB substrate for microwell applications containing 3,3',5,5'-tetramethylbenzidine in a mildly acidic buffer. This substrate is our most sensitive TMB buffer. This sensitive, faster kinetic TMB substrate is approximately 40-fold more sensitive than our most popular substrate, TMB Substrate (ELISA).