Learn More
Seracare Life Sciences Inc TMB SUBSTRATE SOL B 1X100ML
Catalog No. NC0393349
Encompass_Preferred
Most sensitive chromogenic substrate for peroxidase detection, more sensitive than ABTS and OPD. It develops a deep blue color in the presence of peroxidase-labeled conjugates measurable at 650 nm. Turns yellow when stopped by acidification and read at 450 nm. Recommended for ELISA and blotting applications. 2-component format of 3, 3', 5, 5' - Tetramethylbenzidine, simply mix and use. Has longer shelf life than 1-component TMB substrates.
This item is not returnable. View return policy
Catalog No. NC0393349 Supplier Seracare Life Sciences Inc Supplier No. 51200053
Most sensitive chromogenic substrate for peroxidase detection, more sensitive than ABTS and OPD. It develops a deep blue color in the presence of peroxidase-labeled conjugates measurable at 650 nm. Turns yellow when stopped by acidification and read at 450 nm. Recommended for ELISA and blotting applications. 2-component format of 3, 3', 5, 5' - Tetramethylbenzidine, simply mix and use. Has longer shelf life than 1-component TMB substrates.