Learn More
Cayman Chemical TMB (hydrochloride hydrate), Purity: ≥98%, CAS: 207738-08-7, 1 g
SDP
Catalog No. 502896905
Encompass_Preferred
A colorimetric peroxidase substrate; enzymatic oxidation produces a radical cation followed by a diimine product that can be quantified by colorimetric detection at 652 and 450 nm, respectively, as a measure of peroxidase activity; has commonly been used as a substrate for HRP in ELISA and immunohistochemical applications
Catalog No. 50-289-6905 Supplier Cayman Chemical Supplier No. 704501 G
A colorimetric peroxidase substrate; enzymatic oxidation produces a radical cation followed by a diimine product that can be quantified by colorimetric detection at 652 and 450 nm, respectively, as a measure of peroxidase activity; has commonly been used as a substrate for HRP in ELISA and immunohistochemical applications