Learn More
Medchemexpress LLC HY-137175 5mg Medchemexpress, TMBIM6 antagonist-1 CAS:123134-61-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1371755MG
Encompass_Preferred
Medchemexpress, HY-137175 5mg TMBIM6 antagonist-1 CAS:123134-61-2 TMBIM6 antagonist-1, a potential TMBIM6 antagonist, prevents TMBIM6 binding to mTORC2, decreases mTORC2 activity, and also regulates TMBIM6-leaky Ca2+. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0472