Learn More
Bioworld TMB Stop Reagent, 450 nm, 100 mL
Catalog No. 50198884
Encompass_Preferred
TMB Stop ReagentStop reagent is 0.16M sulfuric acid used to stop the ELISA substrate 3,3’,5,5’ - tetramethylbenzidine (TMB) reaction. Horseradish peroxidase (HRP) enzyme conjugates react withTMB and peroxide to produce a blue byproduct having maximum absorbance at 605nm. Adding Stop reagent changes it to yellow color with absorption maximum at 450 nm and stabilizes the color development.Quantity: 100ml
Catalog No. 50-198-884 Supplier Bioworld Supplier No. 215300711
TMB Stop ReagentStop reagent is 0.16M sulfuric acid used to stop the ELISA substrate 3,3’,5,5’ - tetramethylbenzidine (TMB) reaction. Horseradish peroxidase (HRP) enzyme conjugates react withTMB and peroxide to produce a blue byproduct having maximum absorbance at 605nm. Adding Stop reagent changes it to yellow color with absorption maximum at 450 nm and stabilizes the color development.Quantity: 100ml