Learn More
Abcam TMCB, Selective CK2 inhibitor, 10MG
Catalog No. 502707716
Encompass_Preferred
MW 534.82 Da, Purity >98%. CK2 inhibitor (Ki = 0.25 μM), more selective than DMAT. Unlike DMAT, TMCB displays selectivity over PIM1, HIPK2 and DYRK1a (Ki values are 8.65, 15.25 and 11.90 μM, respectively) and a more favourable selectivity profile over a range of other kinases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7716 Supplier Abcam Supplier No. AB12028910MG
MW 534.82 Da, Purity >98%. CK2 inhibitor (Ki = 0.25 μM), more selective than DMAT. Unlike DMAT, TMCB displays selectivity over PIM1, HIPK2 and DYRK1a (Ki values are 8.65, 15.25 and 11.90 μM, respectively) and a more favourable selectivity profile over a range of other kinases.
The product is subject to the following: Abcam Restricted Use Statement