Learn More
Aviva Systems Biology Corp TMCC1 Antibody - middle region (ARP44708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMCC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMCC1. Alternate names: DKFZp686M0169, FLJ42680ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58880