Learn More
Aviva Systems Biology Corp TMCO1 Antibody - C-terminal region (ARP49429_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49429_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMCO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMCO1. Alternate names: HP10122, PCIA3, PNAS-136, RP11-466F5.7, TMCC4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60083