Learn More
Aviva Systems Biology Corp Tmco3 Antibody - middle region (ARP47377_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47377_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tmco3. It was validated on Western Blot. . Alternate names: B230339H12Rik, C87304ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-299