Learn More
Aviva Systems Biology Corp TMCO4 Antibody - C-terminal region (ARP55797_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55797_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMCO4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TMCO4. Alternate names: DKFZp686C23231, RP5-1056L3.6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-083