Learn More
Eurofins Discoverx TMDP, 250 µg. (14-767M)
Supplier:  Eurofins Discoverx 14767M
Encompass
The TMDP product is purified N terminal GST-tagged recombinant TMDP enzyme. This enzyme is full length. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 10 μg of 4.35 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9314