Learn More
Aviva Systems Biology Corp TMED3 Antibody - C-terminal region (ARP46845_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46845_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TMED3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMED3. Alternate names: C15orf22, MGC133022, P24B, p26ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59419