Learn More
Aviva Systems Biology Corp TMED4 Antibody - N-terminal region (ARP41190_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41190_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TMED4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TMED4. Alternate names: HNLF, ERS25ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57754