Learn More
Aviva Systems Biology Corp TMEFF1 Antibody - middle region (ARP45503_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45503_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEFF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEFF1. Alternate names: C9orf2, H7365, TR-1, CT120.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59045