Learn More
Aviva Systems Biology Corp Tmem106b Antibody - C-terminal region (ARP47439_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47439_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tmem106b. It was validated on Western Blot. . Alternate names: 2310036D22Rik, 5830455K21Rik, 6430519M21Rik, AI428776, AI661344ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-305