Learn More
Aviva Systems Biology Corp TMEM106C Antibody - middle region (ARP49796_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49796_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM106C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEM106C. Alternate names: MGC111210, MGC5576ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast
Catalog No. 50-014-60186