Learn More
Aviva Systems Biology Corp TMEM107 Antibody - N-terminal region (ARP50065_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50065_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM107. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM107. Alternate names: GRVS638, MGC10744, PRO1268ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-479