Learn More
Aviva Systems Biology Corp STIMATE Antibody - N-terminal region (ARP55905_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55905_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM110. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM110. Alternate names: DKFZp667E1121, MGC52022ApplicationWB. Species reactivity: Human, Rabbit, Rat
Catalog No. 50-014-61658