Learn More
Aviva Systems Biology Corp TMEM115 Antibody - N-terminal region (ARP46814_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46814_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM115. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM115. Alternate names: PL6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59410