Learn More
Aviva Systems Biology Corp Tmem117 Antibody - C-terminal region (ARP50052_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50052_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tmem117. It was validated on Western Blot. . Alternate names: RGD1562562ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-478