Learn More
Aviva Systems Biology Corp TMEM118 Antibody - N-terminal region (ARP50130_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50130_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM118. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM118. Alternate names: FLJ14627, TMEM118ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53619