Learn More
Aviva Systems Biology Corp TMEM123 Antibody - C-terminal region (ARP50227_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50227_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM123. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMEM123. Alternate names: KCT3, PORIMIN, PORMINApplicationWB. Species reactivity: Human
Catalog No. 50-014-53621