Learn More
Aviva Systems Biology Corp TMEM126B Antibody - middle region (ARP49321_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49321_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM126B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEM126B. Alternate names: HT007, MGC111203ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53581