Learn More
Aviva Systems Biology Corp TMEM127 Antibody - middle region (ARP47363_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47363_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM127. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEM127. Alternate names: FLJ20507, FLJ22257ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59524