Learn More
Aviva Systems Biology Corp Tmem130 Antibody - C-terminal region (ARP37573_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37573_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tmem130. It was validated on Western Blot. Tmem130,transmembrane protein 130,BC067004; MGC157450; MGC157451; C130036G08; Tmem130,transmembrane protein 130,transmembrane protein 130,OTTMUSP00000031094The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: BC067004, C130036G08, MGC157450, MGC157451ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-658