Learn More
Aviva Systems Biology Corp TMEM138 Antibody - N-terminal region (ARP47254_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47254_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM138. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM138. Alternate names: HSPC196, JBTS16ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59500