Learn More
Aviva Systems Biology Corp TMEM139 Antibody - N-terminal region (ARP53159_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53159_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM139. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM139. Alternate names: FLJ90586ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-285-802