Learn More
Aviva Systems Biology Corp TMEM141 Antibody - C-terminal region (ARP60722_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60722_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM141. It was validated on Western Blot. . Alternate names: MGC14141, RP11-216L13.7ApplicationWB. Species reactivity: Cow, Human, Mouse
Catalog No. 50-287-142