Learn More
Aviva Systems Biology Corp TMEM158 Antibody - middle region (ARP47161_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47161_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM158. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEM158. Alternate names: BBP, DKFZp586E1621, RIS1, p40BBPApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-014-59477