Learn More
Aviva Systems Biology Corp TMEM166 Antibody - middle region (ARP58963_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58963_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM166. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TMEM166. Alternate names: FLJ13391, TMEM166ApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-286-543