Learn More
Aviva Systems Biology Corp TMEM168 Antibody - C-terminal region (ARP49730_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49730_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM168. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMEM168. Alternate names: DKFZp564C012, FLJ13576ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60162