Learn More
Aviva Systems Biology Corp TMEM16A Antibody - middle region (ARP42506_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42506_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM16A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEM16A. Alternate names: FLJ10261, ORAOV2, TAOS2, CACC, DOG1, TMEM16A, ANO1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53342