Learn More
Aviva Systems Biology Corp TMEM16C Antibody - C-terminal region (ARP49997_P050) | 100uL
SDP
Catalog No. 5001460235
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM16C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMEM16C. Alternate names: C11orf25, GENX-3947, TMEM16CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60235 Supplier Aviva Systems Biology Corp Supplier No. ARP49997_P050
This is a rabbit polyclonal antibody against TMEM16C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMEM16C. Alternate names: C11orf25, GENX-3947, TMEM16CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat