Learn More
Aviva Systems Biology Corp TMEM173 Antibody - middle region (ARP48041_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48041_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM173. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEM173. Alternate names: FLJ38577, ERIS, MITA, MPYS, NET23, STINGApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-53535