Learn More
Medchemexpress LLC TMEM173 Human N-Hi 250ug
SDP
Catalog No. 5000184287
Encompass_Preferred
Recombinant human TMEM173 (STING) protein expressed in E. coli and supplied with N-terminal SUMO and 6xHis affinity tags. The purified protein is intended for use in biochemical, binding, and immunological assays to investigate innate immune signaling and type I interferon responses. Refer to the product datasheet and COA for storage, handling, and concentration details.
  • Recombinant human protein expressed in E. coli.
  • N-terminal SUMO and 6xHis affinity tags for purification.
  • Purified for biochemical, binding, and immunological assays.
  • Suitable for studies of innate immune signaling and interferon responses.
  • Multiple size options; consult the datasheet for storage and handling.
Catalog No. 50-001-84287 Supplier Medchemexpress LLC Supplier No. HYP70700A250UG
Recombinant human TMEM173 (STING) protein expressed in E. coli and supplied with N-terminal SUMO and 6xHis affinity tags. The purified protein is intended for use in biochemical, binding, and immunological assays to investigate innate immune signaling and type I interferon responses. Refer to the product datasheet and COA for storage, handling, and concentration details.
  • Recombinant human protein expressed in E. coli.
  • N-terminal SUMO and 6xHis affinity tags for purification.
  • Purified for biochemical, binding, and immunological assays.
  • Suitable for studies of innate immune signaling and interferon responses.
  • Multiple size options; consult the datasheet for storage and handling.