Learn More
Medchemexpress LLC TMEM173 Human N-Hi 500ug
SDP
Catalog No. 5000184435
Encompass_Preferred
Recombinant human TMEM173 (STING) protein with an N-terminal SUMO fusion and N-terminal 6xHis affinity tag, expressed in Escherichia coli and supplied as a purified research reagent for biochemical and signaling studies, including investigations of the cGAS-STING pathway.
  • N-terminal SUMO and 6xHis purification tags.
  • Expressed in Escherichia coli for bacterial expression.
  • Supplied as purified recombinant protein for research use.
  • Suitable for biochemical assays and signaling pathway studies.
  • Available in a 500 μg package for assay development and characterization.
Catalog No. 50-001-84435 Supplier Medchemexpress LLC Supplier No. HYP70700A500UG
Recombinant human TMEM173 (STING) protein with an N-terminal SUMO fusion and N-terminal 6xHis affinity tag, expressed in Escherichia coli and supplied as a purified research reagent for biochemical and signaling studies, including investigations of the cGAS-STING pathway.
  • N-terminal SUMO and 6xHis purification tags.
  • Expressed in Escherichia coli for bacterial expression.
  • Supplied as purified recombinant protein for research use.
  • Suitable for biochemical assays and signaling pathway studies.
  • Available in a 500 μg package for assay development and characterization.