Learn More
Medchemexpress LLC TMEM173 Human N-Hi 50ug
SDP
Catalog No. 5000185080
Encompass_Preferred
Recombinant human TMEM173 protein with N-terminal SUMO and 6xHis tags, expressed in E. coli and supplied lyophilized (50 μg). TMEM173 (STING) senses cytoplasmic DNA to activate innate immune signaling and type I interferon production. This preparation is a partial fragment (187 amino acids, ~35.19 kDa), supplied at >95% purity and formulated in 20 mM PB, 150 mM NaCl, pH 7.4.
Catalog No. 50-001-85080 Supplier Medchemexpress LLC Supplier No. HYP70700A50UG
Recombinant human TMEM173 protein with N-terminal SUMO and 6xHis tags, expressed in E. coli and supplied lyophilized (50 μg). TMEM173 (STING) senses cytoplasmic DNA to activate innate immune signaling and type I interferon production. This preparation is a partial fragment (187 amino acids, ~35.19 kDa), supplied at >95% purity and formulated in 20 mM PB, 150 mM NaCl, pH 7.4.