Learn More
Medchemexpress LLC TMEM173 Human N-Hi 5ug | 5ug
SDP
Catalog No. 5000184436
Encompass_Preferred
The TMEM173 protein acts as a promoter of innate immune signaling acting as a sensor of bacterial and viral cytoplasmic DNA ultimately promoting the production of type I interferons (IFN- and IFN- ) This innate immune response is triggered in response to the delivery of non-CpG double-stranded DNA from viruses and bacteria into the cytoplasm TMEM173 Protein Human (N-His-Sumo) is the recombinant human-derived TMEM173 protein expressed by E coli with N-6 His N-SUMO labeled tag
Catalog No. 50-001-84436 Supplier Medchemexpress LLC Supplier No. HYP70700A5UG
The TMEM173 protein acts as a promoter of innate immune signaling acting as a sensor of bacterial and viral cytoplasmic DNA ultimately promoting the production of type I interferons (IFN- and IFN- ) This innate immune response is triggered in response to the delivery of non-CpG double-stranded DNA from viruses and bacteria into the cytoplasm TMEM173 Protein Human (N-His-Sumo) is the recombinant human-derived TMEM173 protein expressed by E coli with N-6 His N-SUMO labeled tag