Learn More
Aviva Systems Biology Corp TMEM176A Antibody - N-terminal region (ARP43538_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43538_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM176A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM176A. Alternate names: GS188, HCA112ApplicationWB. Species reactivity: Human
Catalog No. 50-285-078