Learn More
Aviva Systems Biology Corp TMEM176B Antibody - C-terminal region (ARP42388_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42388_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM176B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TMEM176B. Alternate names: FLJ26014, LR8, MGC110857ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-284-999