Learn More
Aviva Systems Biology Corp MFSD13A Antibody - middle region (ARP49872_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49872_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM180. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TMEM180. Alternate names: C10orf77, FLJ22529, bA18I14.8, RP11-18I14.8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-459